accessibility menu, dialog, popup

UserName

CAS RN 19932-85-5

BrHNOO

CAS RN 19932-85-5

IUPAC Name: 6-bromo-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 2-benzoxazolinone, 6-bromo-
Molecular Weight (g/mol): 214.02
Molecular Formula: C7H4BrNO2
InChi Key: DDNKJFBQMQOIKI-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)