accessibility menu, dialog, popup

UserName

H3C(S)NH2OHO

CAS RN 1991-87-3

IUPAC Name: (2S)-2-amino-3-(4-methylphenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(4-methylphenyl)propanoic acid
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: DQLHSFUMICQIMB-VIFPVBQESA-N
SMILES: CC1=CC=C(C[C@H](N)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results