CAS RN 199006-54-7
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methylphenyl)propanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid
Molecular Weight (g/mol):
401.46
Molecular Formula:
C25H23NO4
InChi Key:
UXLHLZHGQPDMJQ-QHCPKHFHSA-N
SMILES:
CC1=CC=C(C[C@H](NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
1
–
1
of
1
results
1
N-Fmoc-4-methyl-L-phenylalanine, 95%
CAS: 199006-54-7 Molecular Formula: C25H23NO4 Molecular Weight (g/mol): 401.462 MDL Number: MFCD00270197 InChI Key: UXLHLZHGQPDMJQ-QHCPKHFHSA-N Synonym: fmoc-phe 4-me-oh,fmoc-l-4-methylphe,fmoc-4-methyl-l-phenylalanine,fmoc-l-4-methylphenylalanine,fmoc-4-methy-l-phenylalanine,fmoc-l-phe 4-me-oh,fmoc-p-me-phe-oh,n-fmoc-4-methyl-l-phenylalanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid,phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl PubChem CID: 7129826 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid SMILES: CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24