accessibility menu, dialog, popup

UserName

H2NNOOHOO

CAS RN 19883-74-0

IUPAC Name: 2-amino-3-(3-nitrophenyl)propanoic acid
Synonyms: 2-amino-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: YTHDRUZHNYKZGF-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)