accessibility menu, dialog, popup

UserName

CAS RN 1988-88-1

CH3H3CCH3HOH3CH3CCH3N

CAS RN 1988-88-1

IUPAC Name: 3,5-di-tert-butyl-4-hydroxybenzonitrile
Synonyms: 3,5-di-tert-butyl-4-hydroxybenzonitrile
Molecular Weight (g/mol): 231.34
Molecular Formula: C15H21NO
InChi Key: AKXIIOLURNATOC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results