accessibility menu, dialog, popup

UserName

CAS RN 19830-09-2

CH3OCH3OOOO

CAS RN 19830-09-2

IUPAC Name: 4-acetyl-4-methylheptanedioate
Synonyms: 4-acetyl-4-methylheptanedioate
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: LYBQMKJABQTTFJ-UHFFFAOYSA-L
SMILES: CC(=O)C(C)(CCC([O-])=O)CCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results