accessibility menu, dialog, popup

UserName

CAS RN 1982-47-4

H3CNCH3ONHOCl

CAS RN 1982-47-4

IUPAC Name: 1-[4-(4-chlorophenoxy)phenyl]-3,3-dimethylurea
Synonyms: chloroxuron
Molecular Weight (g/mol): 290.75
Molecular Formula: C15H15ClN2O2
InChi Key: IVUXTESCPZUGJC-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results