accessibility menu, dialog, popup

UserName

CH3H3COHCH3H3C

CAS RN 19781-13-6

IUPAC Name: 4,7-dimethyloctan-4-ol
Synonyms: 4,7-dimethyloctan-4-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: OTISWHNJCUMBFI-UHFFFAOYNA-N
SMILES: CCCC(C)(O)CCC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results