accessibility menu, dialog, popup

UserName

CAS RN 19780-78-0

H3C(S)CH3(R)CH3OH

CAS RN 19780-78-0

IUPAC Name: (2R,4S)-4-ethyloctan-2-ol
Synonyms: (2R,4S)-4-ethyloctan-2-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: GVKBTEISGUWZEJ-ZJUUUORDSA-N
SMILES: CCCC[C@H](CC)C[C@@H](C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results