accessibility menu, dialog, popup

UserName

CAS RN 19780-66-6

H3CCH3CH3H2C

CAS RN 19780-66-6

IUPAC Name: 3-ethyl-2-methylpent-1-ene
Synonyms: 1-pentene, 3-ethyl-2-methyl-
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: HPHHYSWOBXEIRG-UHFFFAOYSA-N
SMILES: CCC(CC)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results