accessibility menu, dialog, popup

UserName

OClSFFFFF

CAS RN 197384-98-8

IUPAC Name: 4-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Synonyms: 4-(pentafluoro-λ⁶-sulfanyl)benzoyl chloride
Molecular Weight (g/mol): 266.61
Molecular Formula: C7H4ClF5OS
InChi Key: NNFDYUZNKYWPGP-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results