accessibility menu, dialog, popup

UserName

CAS RN 1972-28-7

IUPAC Name: N-[(ethoxycarbonyl)imino]ethoxyformamide
Synonyms: diethyl azodicarboxylate
Molecular Weight (g/mol): 174.16
Molecular Formula: C6H10N2O4
InChi Key: FAMRKDQNMBBFBR-UHFFFAOYSA-N
SMILES: CCOC(=O)N=NC(=O)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results