accessibility menu, dialog, popup

UserName

CAS RN 196961-43-0

H3COHOOHCH3H3CH3CCH3(E/Z)

CAS RN 196961-43-0

IUPAC Name: 3-[4-hydroxy-3-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)phenyl]prop-2-enoic acid
Synonyms: 3-[4-hydroxy-3-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 364.49
Molecular Formula: C24H28O3
InChi Key: DYLLZSVPAUUSSB-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C=C1C1=C(O)C=CC(C=CC(O)=O)=C1)C(C)(C)CCC2(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results