CAS RN 19666-76-3
CAS RN 19666-76-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Panaxadiol
MedChemExpress Panaxadiol is found in the roots of Panax ginseng, activities neuroprotective and anti-tumour functions. Panaxadiol inhibits programmed cell death-ligand 1(PD-L1) expression and tumour proliferation.
2
MedChemExpress Panaxadiol
MedChemExpress Panaxadiol is found in the roots of Panax ginseng, activities neuroprotective and anti-tumour functions. Panaxadiol inhibits programmed cell death-ligand 1(PD-L1) expression and tumour proliferation.