accessibility menu, dialog, popup

UserName

CAS RN 195136-46-0

IUPAC Name: 1-(chloromethyl)-2,4-bis(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-2,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 262.58
Molecular Formula: C9H5ClF6
InChi Key: DPKQSESRNWBUFZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CCl)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results