accessibility menu, dialog, popup

UserName

CAS RN 19500-95-9

H3COOOOOCH3

CAS RN 19500-95-9

IUPAC Name: 2-methoxyacetyl 2-methoxyacetate
Synonyms: 2-methoxyacetyl 2-methoxyacetate
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: PEHFQQWAINXOQG-UHFFFAOYSA-N
SMILES: COCC(=O)OC(=O)COC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results