accessibility menu, dialog, popup

UserName

CAS RN 194491-31-1

NNOOOOOOOONaNaNaNa

CAS RN 194491-31-1

IUPAC Name: tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate hydrate
Synonyms: tetrasodium ion(4-) edta hydrate
Molecular Weight (g/mol): 398.19
Molecular Formula: C10H14N2Na4O9
InChi Key: GXFFSKMBMYHTAE-UHFFFAOYSA-J
SMILES: O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)