accessibility menu, dialog, popup

UserName

CAS RN 194163-91-2

CH3H3CCH3OONOOH3CN

CAS RN 194163-91-2

IUPAC Name: 1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 254.29
Molecular Formula: C12H18N2O4
InChi Key: GHLKJIQVORAVHE-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results