accessibility menu, dialog, popup

UserName

CAS RN 19406-86-1

CH3H2NNH2O

CAS RN 19406-86-1

IUPAC Name: 3-amino-4-methylbenzamide
Synonyms: 3-amino-4-methylbenzamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: VYBKAZXQKUFAHG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N)C(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)