accessibility menu, dialog, popup

UserName

CAS RN 19404-18-3

H3CSCl

CAS RN 19404-18-3

IUPAC Name: 5-chloro-3-methyl-1-benzothiophene
Synonyms: 5-chloro-3-methyl-1-benzothiophene
Molecular Weight (g/mol): 182.67
Molecular Formula: C9H7ClS
InChi Key: UWDQVEPXORTQFO-UHFFFAOYSA-N
SMILES: CC1=CSC2=CC=C(Cl)C=C12
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)