accessibility menu, dialog, popup

UserName

NaNaNaOONNSOOOONNSOOO(E/Z)

CAS RN 1934-21-0

IUPAC Name: trisodium 5-oxo-1-(4-sulfonatophenyl)-4-[2-(4-sulfonatophenyl)diazen-1-yl]-4,5-dihydro-1H-pyrazole-3-carboxylate
Synonyms: tartrazine
Molecular Weight (g/mol): 534.36
Molecular Formula: C16H9N4Na3O9S2
InChi Key: UJMBCXLDXJUMFB-UHFFFAOYNA-K
SMILES: [Na+].[Na+].[Na+].[O-]C(=O)C1=NN(C(=O)C1N=NC1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)