accessibility menu, dialog, popup

UserName

ClNClClCl

CAS RN 1929-82-4

IUPAC Name: 2-chloro-6-(trichloromethyl)pyridine
Synonyms: nitrapyrin
Molecular Weight (g/mol): 230.9
Molecular Formula: C6H3Cl4N
InChi Key: DCUJJWWUNKIJPH-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results