accessibility menu, dialog, popup

UserName

CAS RN 192863-37-9

IUPAC Name: potassium trifluoro(thiophen-3-yl)boranuide
Synonyms: potassium trifluoro(thiophen-3-yl)boranuide
Molecular Weight (g/mol): 190.03
Molecular Formula: C4H3BF3KS
InChi Key: ULOOPHJJEPUNNH-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results