accessibility menu, dialog, popup

UserName

CAS RN 192804-36-7

HOBOHNHOO

CAS RN 192804-36-7

IUPAC Name: (4-{[(benzyloxy)carbonyl]amino}phenyl)boronic acid
Synonyms: 4-{[(benzyloxy)carbonyl]amino}phenylboronic acid
Molecular Weight (g/mol): 271.08
Molecular Formula: C14H14BNO4
InChi Key: KYCQBMOJDXVNIC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(NC(=O)OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results