accessibility menu, dialog, popup

UserName

CAS RN 19241-24-8

CH3H3CCH3NCS

CAS RN 19241-24-8

IUPAC Name: 1-tert-butyl-4-isothiocyanatobenzene
Synonyms: 1-tert-butyl-4-isothiocyanatobenzene
Molecular Weight (g/mol): 191.29
Molecular Formula: C11H13NS
InChi Key: OCGNNCBNRBTUCG-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results