CAS RN 192374-14-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S,S)-Hydroxy Bupropion, TRC
CAS: 192374-14-4 Molecular Formula: C13 H18 Cl N O2 . Cl H Molecular Weight (g/mol): 292.2 Synonym: (2S,3S)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol,Radafaxine IUPAC Name: (2S,3S)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol;hydrochloride SMILES: Cl.C[C@@H]1NC(C)(C)CO[C@@]1(O)c2cccc(Cl)c2