accessibility menu, dialog, popup

UserName

CAS RN 192323-12-9

H3COOOCH3SOOCl

CAS RN 192323-12-9

IUPAC Name: methyl 3-(chlorosulfonyl)-4-methoxybenzoate
Synonyms: methyl 3-(chlorosulfonyl)-4-methoxybenzoate
Molecular Weight (g/mol): 264.68
Molecular Formula: C9H9ClO5S
InChi Key: FKAIQWMEWVOGDP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results