accessibility menu, dialog, popup

UserName

CAS RN 1918-77-0

OHOS

CAS RN 1918-77-0

IUPAC Name: 2-(thiophen-2-yl)acetic acid
Synonyms: thiolacetic acid 2-thiopheneacetic acid
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: SMJRBWINMFUUDS-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)