accessibility menu, dialog, popup

UserName

CAS RN 19179-31-8

H3COOCH3N

CAS RN 19179-31-8

IUPAC Name: 3,5-dimethoxybenzonitrile
Synonyms: benzonitrile, 3,5-dimethoxy-
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: NVTHWSJNXVDIKR-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results