accessibility menu, dialog, popup

UserName

H3CNBrOO

CAS RN 19165-25-4

IUPAC Name: 6-bromo-2-methyl-4H-3,1-benzoxazin-4-one
Synonyms: 6-bromo-2-methyl-3,1-benzoxazin-4-one
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: JCRULBKTDUOWMP-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(Br)C=C2C(=O)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results