accessibility menu, dialog, popup

UserName

KKBrIrBrBrBrBrBr

CAS RN 19121-78-9

IUPAC Name: dipotassium hexabromoiridiumdiuide
Synonyms: dipotassium hexabromoiridiumdiuide
Molecular Weight (g/mol): 749.84
Molecular Formula: Br6IrK2
InChi Key: RSDYABYRTHADQA-UHFFFAOYSA-H
SMILES: [K+].[K+].Br[Ir--](Br)(Br)(Br)(Br)Br
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results