accessibility menu, dialog, popup

UserName

CAS RN 19093-34-6

H3CSOOHO

CAS RN 19093-34-6

IUPAC Name: 2-methanesulfinylbenzoic acid
Synonyms: 2-methanesulfinylbenzoic acid
Molecular Weight (g/mol): 184.21
Molecular Formula: C8H8O3S
InChi Key: JVYFTEBQXOBPAB-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results