accessibility menu, dialog, popup

UserName

CAS RN 19049-40-2

BeBeBeBeOCH3OOCH3OOCH3OOCH3OOCH3OOCH3OO

CAS RN 19049-40-2

IUPAC Name: tetraberyllium(2+) hexaacetate oxidandiide
Synonyms: basic beryllium acetate
Molecular Weight (g/mol): 406.31
Molecular Formula: C12H18Be4O13
InChi Key: RKARILOACIIVIJ-UHFFFAOYSA-H
SMILES: [Be++].[Be++].[Be++].[Be++].[O--].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results