accessibility menu, dialog, popup

UserName

BOOBOOH2OBa

CAS RN 19004-06-9

IUPAC Name: barium(2+) hydrate dioxoborinate
Synonyms: barium(2+) hydrate dioxoborinate
Molecular Weight (g/mol): 240.96
Molecular Formula: B2BaH2O5
InChi Key: JXOXZJCHHKIXMI-UHFFFAOYSA-N
SMILES: O.[Ba++].[O-]B=O.[O-]B=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results