accessibility menu, dialog, popup

UserName

CAS RN 18996-35-5

NaOHOHOOHOOO

CAS RN 18996-35-5

IUPAC Name: sodium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Synonyms: monosodium citrate
Molecular Weight (g/mol): 214.11
Molecular Formula: C6H7NaO7
InChi Key: HWPKGOGLCKPRLZ-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)CC(O)(CC(O)=O)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Grade 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)