accessibility menu, dialog, popup

UserName

CAS RN 18962-05-5

CH3H3COO

CAS RN 18962-05-5

IUPAC Name: 4-(propan-2-yloxy)benzaldehyde
Synonyms: 4-isopropoxybenzaldehyde
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: WDANSDASCKBVKH-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(C=O)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results