accessibility menu, dialog, popup

UserName

CAS RN 1891-95-8

IUPAC Name: 3,5-dichloro-4-hydroxybenzonitrile
Synonyms: 3,5-dichloro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: YRSSHOVRSMQULE-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(C=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results