accessibility menu, dialog, popup

UserName

CAS RN 188813-02-7

FBrO

CAS RN 188813-02-7

IUPAC Name: 3-bromo-5-fluorobenzaldehyde
Synonyms: 3-bromo-5-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: MEFQRXHVMJPOKZ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(C=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results