accessibility menu, dialog, popup

UserName

CAS RN 1887-60-1

CH3NSNCH3

CAS RN 1887-60-1

IUPAC Name: 5,6-dimethyl-2,1,3-benzothiadiazole
Synonyms: 5,6-dimethyl-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: QQCMLCGOLBUDJF-UHFFFAOYSA-N
SMILES: CC1=CC2=NSN=C2C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results