accessibility menu, dialog, popup

UserName

CAS RN 188527-21-1

IUPAC Name: 1-[(benzyloxy)carbonyl]pyrrolidine-3-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 249.27
Molecular Formula: C13H15NO4
InChi Key: JSASVUTVTRNJHA-UHFFFAOYNA-N
SMILES: OC(=O)C1CCN(C1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results