accessibility menu, dialog, popup

UserName

CAS RN 1885-29-6

NH2N

CAS RN 1885-29-6

IUPAC Name: 2-aminobenzonitrile
Synonyms: benzonitrile, 2-amino-
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: HLCPWBZNUKCSBN-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C#N
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)