CAS RN 188425-85-6
IUPAC Name:
2-chloro-N-{4'-chloro-[1,1'-biphenyl]-2-yl}pyridine-3-carboxamide
Synonyms:
boscalid
Molecular Weight (g/mol):
343.21
Molecular Formula:
C18H12Cl2N2O
InChi Key:
WYEMLYFITZORAB-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=CC=CC=C1NC(=O)C1=CC=CN=C1Cl
1
–
3
of
3
results
1
Boscalid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Boscalid, TRC
CAS: 188425-85-6 Molecular Formula: C18 H12 Cl2 N2 O Molecular Weight (g/mol): 343.21 Synonym: 2-Chloro-N-(4'-chloro[1,1'-biphenyl]-2-yl)-3-pyridinecarboxamide,2-Chloro-N-(4'-chlorobiphenyl-2-yl)nicotinamide,BAS 510F,Boscalid,Cantus,Emerald,Emerald (fungicide),Endura,F 510,Filan,Nicobifen IUPAC Name: 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide SMILES: Clc1ccc(cc1)c2ccccc2NC(=O)c3cccnc3Cl
3
Boscalid, Fluka™
Analytical standard, reference material