accessibility menu, dialog, popup

UserName

CAS RN 18829-56-6

H3CO(E)

CAS RN 18829-56-6

IUPAC Name: (2E)-non-2-enal
Synonyms: trans-non-2-enal
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: BSAIUMLZVGUGKX-BQYQJAHWSA-N
SMILES: CCCCCC\C=C\C=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)