CAS RN 18807-73-3
IUPAC Name:
benzyl N-(4-aminobutyl)carbamate hydrochloride
Synonyms:
benzyl N-(4-aminobutyl)carbamate hydrochloride
Molecular Weight (g/mol):
258.75
Molecular Formula:
C12H19ClN2O2
InChi Key:
ZVNNCIIFBSRHFE-UHFFFAOYSA-N
SMILES:
Cl.NCCCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-CBZ-1,4-Diaminobutane HCl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.