accessibility menu, dialog, popup

UserName

CH3NNOH3COOH

CAS RN 187998-64-7

IUPAC Name: 1-(4-methoxyphenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 232.24
Molecular Formula: C12H12N2O3
InChi Key: JPFGKGZYCXLEGQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1N=CC(C(O)=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results