accessibility menu, dialog, popup

UserName

CAS RN 1879-56-7

OOOBr

CAS RN 1879-56-7

IUPAC Name: 2-(2-bromophenoxy)acetate
Synonyms: 2-(2-bromophenoxy)acetate
Molecular Weight (g/mol): 230.04
Molecular Formula: C8H6BrO3
InChi Key: AZYODYPUWJPKOI-UHFFFAOYSA-M
SMILES: [O-]C(=O)COC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results