missing translation for 'onlineSavingsMsg'
Learn More

o-Bromophenoxyacetic acid, TRC

Product Code. 30330265
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330265 Supplier Toronto Research Chemicals Supplier No. TRCB687233100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Bromophenoxyacetic acid
CAS 1879-56-7
Molecular Formula C8H7BrO3
Quantity 100 mg
InChI Formula InChI=1S/C8H7BrO3/c9-6-3-1-2-4-7(6)12-5-8(10)11/h1-4H,5H2,(H,10,11)
SMILES OC(=O)COc1ccccc1Br
IUPAC Name 2-(2-bromophenoxy)acetic acid
Molecular Weight (g/mol) 231.04
Formula Weight 229.9579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.