accessibility menu, dialog, popup

UserName

OHOCl

CAS RN 1878-66-6

IUPAC Name: 2-(4-chlorophenyl)acetic acid
Synonyms: (4-chlorophenyl)acetic acid C8H6ClO2 P-chlorophenylacetic acid C8H7ClO2
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: CDPKJZJVTHSESZ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)