CAS RN 186817-57-2
CAS RN 186817-57-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hexakis(2,2-difluoroethoxy)phosphazene, TRC
CAS: 186817-57-2 Molecular Formula: C12H18F12N3O6P3 Molecular Weight (g/mol): 621.19 Synonym: Hexakis(1H,1H,2H-perfluoroethoxy)phosphazene;2,2,4,4,6,6-Hexakis(2,2-difluoroethoxy)-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-triazatriphosphorine IUPAC Name: 2,2,4,4,6,6-hexakis(2,2-difluoroethoxy)-1,3,5-triaza-2λ^{5},4λ^{5},6λ^{5}-triphosphacyclohexa-1,3,5-triene SMILES: FC(F)COp1(OCC(F)F)np(OCC(F)F)(OCC(F)F)np(OCC(F)F)(OCC(F)F)n1