accessibility menu, dialog, popup

UserName

CAS RN 18643-08-8

H3CSiCH3CH3Cl

CAS RN 18643-08-8

IUPAC Name: chlorodimethyloctadecylsilane
Synonyms: chlorodimethyloctadecylsilane
Molecular Weight (g/mol): 347.1
Molecular Formula: C20H43ClSi
InChi Key: GZGREZWGCWVAEE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results